Gene Transcripts
- Gene Symbol
- A930009A15Rik
- Entrez Gene ID
- 77798
- Gene Name
- RIKEN cDNA A930009A15 gene
- Gene Aliases
- Chromosome Location
- Chr. 10: 115569809 - 115582454 on Build GRCm38
- UniGene ID
- Mm.446685
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020834.1 |
- |
5 |
970 |
|
View Full Product Details