Gene Transcripts
- Gene Symbol
- C330026H20Rik
- Entrez Gene ID
- 109358
- Gene Name
- RIKEN cDNA C330026H20 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.35204
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037477.1 |
- |
1 |
2573 |
|
AK082813.1 |
- |
2 |
236 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB651998.1 |
- |
2 |
236 |
|
Chr. 19 - 41005779 - 41006574 [-] |
View Full Product Details