Gene Transcripts
- Gene Symbol
- Gm16675
- Entrez Gene ID
- 100503498
- Gene Name
- predicted gene, 16675
- Gene Aliases
- Chromosome Location
- Chr. 8: 46730970 - 46739515 on Build GRCm38
- UniGene ID
- Mm.187358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045750.1 |
|
3 |
581 |
|
GenBank mRNA |
AK144632.1 |
- |
3 |
582 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY734040.1 |
- |
3 |
581 |
|
Chr. 8 - 46731564 - 46739515 [-] |
View Full Product Details