Gene Transcripts
- Gene Symbol
- 2900063K03Rik
- Entrez Gene ID
- 72975
- Gene Name
- RIKEN cDNA 2900063K03 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013743.1 |
- |
1 |
710 |
|
View Full Product Details