Gene Transcripts
- Gene Symbol
- 4930448F12Rik
- Entrez Gene ID
- 73992
- Gene Name
- RIKEN cDNA 4930448F12 gene
- Gene Aliases
- Chromosome Location
- Chr. 13: 18084947 - 18100202 on Build GRCm38
- UniGene ID
- Mm.159110
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046032.1 |
|
2 |
233 |
|
GenBank mRNA |
AK015417.1 |
- |
2 |
234 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714966.1 |
- |
2 |
234 |
|
Chr. 13 - 18084936 - 18100202 [-] |
View Full Product Details