Gene Transcripts
- Gene Symbol
- 1700113A16Rik
- Entrez Gene ID
- 76642
- Gene Name
- RIKEN cDNA 1700113A16 gene
- Gene Aliases
- 2210408E11Rik, AA408983
- Chromosome Location
- Chr. 3: 88171560 - 88177785 on Build GRCm38
- UniGene ID
- Mm.424420
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045997.1 |
|
3 |
931 |
|
GenBank mRNA |
AK008852.1 |
- |
3 |
929 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG075424.2 |
- |
1 |
474 |
|
Chr. 3 - 88171555 - 88172085 [-] |
GenBankmRNA |
CK341655.1 |
- |
1 |
478 |
|
Chr. 3 - 88171444 - 88172090 [-] |
GenBankmRNA |
CD742364.1 |
- |
1 |
496 |
|
Chr. 3 - 88171343 - 88172091 [-] |
GenBankmRNA |
BI853643.1 |
- |
4 |
724 |
|
Chr. 3 - 88171556 - 88177692 [-] |
View Full Product Details