Gene Transcripts
- Gene Symbol
- 4930519K11Rik
- Entrez Gene ID
- 78019
- Gene Name
- RIKEN cDNA 4930519K11 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.243845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019684.1 |
- |
3 |
322 |
|
BY715389.1 |
- |
3 |
322 |
|
View Full Product Details