Gene Transcripts
- Gene Symbol
- 1810059C17Rik
- Entrez Gene ID
- 69818
- Gene Name
- RIKEN cDNA 1810059C17 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.437204
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007903.1 |
- |
3 |
439 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707906.1 |
- |
3 |
440 |
|
Chr. 2 - 18054193 - 18056613 [-] |
View Full Product Details