Gene Transcripts
- Gene Symbol
- Gimap5
- Entrez Gene ID
- 317757
- Gene Name
- GTPase, IMAP family member 5
- Gene Aliases
- D630024P16, E230026N22Rik
- Chromosome Location
- Chr. 6: 48746137 - 48755235 on Build GRCm38
- UniGene ID
- Mm.455917
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054194.1 |
- |
1 |
2208 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB551287.2 |
- |
1 |
526 |
|
Chr. 6 - 48747980 - 48748622 [+] |
View Full Product Details