Gene Transcripts
- Gene Symbol
- 9130604C24Rik
- Entrez Gene ID
- 77707
- Gene Name
- RIKEN cDNA 9130604C24 gene
- Gene Aliases
- AW554730
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.393410
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020298.1 |
- |
1 |
187 |
|
View Full Product Details