Gene Transcripts
- Gene Symbol
- Gm3916
- Entrez Gene ID
- 100042588
- Gene Name
- predicted gene 3916
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.74650
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086006.1 |
- |
1 |
54 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB662335.1 |
- |
1 |
51 |
|
Chr. 19 - 27260732 - 27262321 [-] |
View Full Product Details