Gene Transcripts
- Gene Symbol
- LOC101927604
- Entrez Gene ID
- 101927604
- Gene Name
- uncharacterized LOC101927604
- Gene Aliases
- Chromosome Location
- Chr. 1: 230874846 - 230879015 on Build GRCh38
- UniGene ID
- Hs.638929
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110681.1 |
|
2 |
359 |
|
GenBank mRNA |
BC032911.1 |
- |
2 |
364 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN292297.1 |
- |
2 |
294 |
|
Chr. 1 - 230874911 - 230875469 [+] |
GENCODE |
ENST00000439341.1 |
- |
2 |
359 |
|
Chr. 1 - 230874846 - 230879015 [+] |
GenBankmRNA |
BI459188.1 |
- |
2 |
362 |
|
Chr. 1 - 230874846 - 230879017 [+] |
GenBankmRNA |
AA992950.1 |
- |
2 |
307 |
|
Chr. 1 - 230874896 - 230875413 [+] |
View Full Product Details