Gene Transcripts
- Gene Symbol
- KCCAT211
- Entrez Gene ID
- 102724550
- Gene Name
- renal clear cell carcinoma-associated transcript 211
- Gene Aliases
- Chromosome Location
- Chr. 3: 177816865 - 177899224 on Build GRCh38
- UniGene ID
- Hs.570526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110826.1 |
|
3 |
745 |
|
GenBank mRNA |
AK056252.1 |
- |
3 |
746 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000436078.5 |
- |
3 |
745 |
|
Chr. 3 - 177816865 - 177899224 [+] |
View Full Product Details