Gene Transcripts
- Gene Symbol
- LINC00911
- Entrez Gene ID
- 100996280
- Gene Name
- long intergenic non-protein coding RNA 911
- Gene Aliases
- Chromosome Location
- Chr. 14: 85393879 - 85420074 on Build GRCh38
- UniGene ID
- Hs.135735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102737.1 |
|
3 |
259 |
|
NR_102738.1 |
|
2 |
149 |
|
GenBank mRNA |
AK131026.1 |
- |
3 |
259 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555272.1 |
- |
3 |
259 |
|
Chr. 14 - 85393879 - 85420074 [+] |
View Full Product Details