Gene Transcripts
- Gene Symbol
- 1700010H22Rik
- Entrez Gene ID
- 75500
- Gene Name
- RIKEN cDNA 1700010H22 gene
- Gene Aliases
- 4930440M09Rik
- Chromosome Location
- Chr. 5: 98562243 - 98566520 on Build GRCm38
- UniGene ID
- Mm.79012
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005827.1 |
- |
|
210 |
|
AK015350.1 |
- |
1 |
288 |
|
View Full Product Details