Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293560.1 | NP_001280489.1 | 12 | 1313 | |
NM_173763.4 | NP_776124.1 | 12 | 1317 | ||
GenBank mRNA | AF363737.1 | - | 12 | 1287 | |
AK049569.1 | - | 12 | 1286 | ||
AK145623.1 | - | 12 | 1311 | ||
BC037135.1 | - | 8 | 894 | ||
BC085287.1 | - | 13 | 1410 | ||
BC131942.1 | - | 11 | 1102 | 40130219 | |
BC132615.1 | - | 11 | 1102 | 40130892 |