Gene Transcripts
- Gene Symbol
- BC061212
- Entrez Gene ID
- 381724
- Gene Name
- cDNA sequence BC061212
- Gene Aliases
- Chromosome Location
- Chr. 5: 94066454 - 94070545 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
- Gene Symbol
- A430089I19Rik
- Entrez Gene ID
- 331195
- Gene Name
- RIKEN cDNA A430089I19 gene
- Gene Aliases
- Gm16522
- Chromosome Location
- Chr. 5: 94302519 - 94306796 on Build GRCm38
- UniGene ID
- Mm.26025
- Species
- Mus musculus
- Gene Symbol
- BC080696
- Entrez Gene ID
- 665521
- Gene Name
- cDNA sequence BC080696
- Gene Aliases
- Chromosome Location
- Chr. 5: 94434131 - 94436940 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC080696.1 |
- |
2 |
521 |
|
- Gene Symbol
- Gm7978
- Entrez Gene ID
- 666203
- Gene Name
- predicted gene 7978
- Gene Aliases
- EG666203
- Chromosome Location
- Chr. 5: 95732092 - 95736173 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
- Gene Symbol
- LOC545763
- Entrez Gene ID
- 545763
- Gene Name
- uncharacterized LOC545763
- Gene Aliases
- Chromosome Location
- Chr. 5|NT_187059: 910224 - 914380 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CN677185.1 |
- |
2 |
491 |
|
View Full Product Details