Gene Transcripts
- Gene Symbol
- Gm10340
- Entrez Gene ID
- 100040697
- Gene Name
- predicted gene 10340
- Gene Aliases
- ENSMUSG00000071613
- Chromosome Location
- Chr. 14: 3130678 - 3140553 on Build GRCm38
- UniGene ID
- Mm.429277
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651812.1 |
- |
1 |
112 |
|
EF651814.1 |
- |
1 |
112 |
|
- Gene Symbol
- Gm10406
- Entrez Gene ID
- 100038847
- Gene Name
- predicted gene 10406
- Gene Aliases
- ENSMUSG00000072735
- Chromosome Location
- Chr. 14: 7006115 - 7028139 on Build GRCm38
- UniGene ID
- Mm.443041
- Species
- Mus musculus
- Gene Symbol
- Gm10413
- Entrez Gene ID
- 100041774
- Gene Name
- predicted gene 10413
- Gene Aliases
- 100038846, 1700001E04Rik, ENSMUSG00000072749, Gm1973
- Chromosome Location
- Chr. 14: 3395093 - 3403534 on Build GRCm38
- UniGene ID
- Mm.360540
- Species
- Mus musculus
- Gene Symbol
- Gm2237
- Entrez Gene ID
- 100039441
- Gene Name
- predicted gene 2237
- Gene Aliases
- ENSMUSG00000079354
- Chromosome Location
- Chr. 14: 19563750 - 19584066 on Build GRCm38
- UniGene ID
- Mm.396155
- Species
- Mus musculus
- Gene Symbol
- Gm2897
- Entrez Gene ID
- 100040671
- Gene Name
- predicted gene 2897
- Gene Aliases
- ENSMUSG00000079410
- Chromosome Location
- Chr. 14: 3049285 - 3085663 on Build GRCm38
- UniGene ID
- Mm.360540
- Species
- Mus musculus
- Gene Symbol
- Gm3002
- Entrez Gene ID
- 100040852
- Gene Name
- alpha-takusan pseudogene
- Gene Aliases
- ENSMUSG00000079399
- Chromosome Location
- Chr. 14: 3814913 - 3830683 on Build GRCm38
- UniGene ID
- Mm.483675
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC080297.1 |
- |
2 |
3745 |
|
- Gene Symbol
- Gm3020
- Entrez Gene ID
- 100040880
- Gene Name
- predicted gene 3020
- Gene Aliases
- ENSMUSG00000079402
- Chromosome Location
- Chr. 14: 3652030 - 3672140 on Build GRCm38
- UniGene ID
- Mm.444311
- Species
- Mus musculus
- Gene Symbol
- Gm3095
- Entrez Gene ID
- 100041012
- Gene Name
- predicted gene 3095
- Gene Aliases
- Chromosome Location
- Chr. 14: 3947711 - 3969145 on Build GRCm38
- UniGene ID
- Mm.460754
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651837.1 |
- |
|
112 |
|
- Gene Symbol
- Gm3173
- Entrez Gene ID
- 100169868
- Gene Name
- predicted gene 3173
- Gene Aliases
- 100041162
- Chromosome Location
- Chr. 14: 4498863 - 4519452 on Build GRCm38
- UniGene ID
- Mm.460196
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651801.1 |
- |
|
112 |
|
- Gene Symbol
- Gm3194
- Entrez Gene ID
- 100042054
- Gene Name
- predicted gene 3194
- Gene Aliases
- ENSMUSG00000079375
- Chromosome Location
- Chr. 14: 5410666 - 5460410 on Build GRCm38
- UniGene ID
- Mm.443041
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651800.1 |
- |
1 |
112 |
|
EF651826.1 |
- |
|
61 |
|
EF651841.1 |
- |
|
61 |
|
- Gene Symbol
- Gm3264
- Entrez Gene ID
- 100041306
- Gene Name
- predicted gene 3264
- Gene Aliases
- Chromosome Location
- Chr. 14: 4853964 - 4875851 on Build GRCm38
- UniGene ID
- Mm.396155
- Species
- Mus musculus
- Gene Symbol
- Gm3317
- Entrez Gene ID
- 666329
- Gene Name
- predicted gene 3317
- Gene Aliases
- 100041401, 100041747
- Chromosome Location
- Chr. 14: 5148982 - 5170292 on Build GRCm38
- UniGene ID
- Mm.429277
- Species
- Mus musculus
- Gene Symbol
- Gm3373
- Entrez Gene ID
- 100036568
- Gene Name
- predicted gene 3373
- Gene Aliases
- 100041501, Gm17143
- Chromosome Location
- Chr. 14: 5719437 - 5880598 on Build GRCm38
- UniGene ID
- Mm.483651
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC040090.1 |
- |
|
668 |
|
BC096548.1 |
- |
|
2153 |
|
- Gene Symbol
- Gm3383
- Entrez Gene ID
- 100041515
- Gene Name
- predicted gene 3383
- Gene Aliases
- ENSMUSG00000079376
- Chromosome Location
- Chr. 14: 5763194 - 5814642 on Build GRCm38
- UniGene ID
- Mm.443041
- Species
- Mus musculus
- Gene Symbol
- Gm3488
- Entrez Gene ID
- 100041735
- Gene Name
- predicted gene, 3488
- Gene Aliases
- 100041747, Gm3494
- Chromosome Location
- Chr. 14: 5501623 - 5522929 on Build GRCm38
- UniGene ID
- Mm.464176
- Species
- Mus musculus
- Gene Symbol
- Gm3500
- Entrez Gene ID
- 100041678
- Gene Name
- predicted gene 3500
- Gene Aliases
- 100041758, Gm3401
- Chromosome Location
- Chr. 14: 5365792 - 5389224 on Build GRCm38
- UniGene ID
- Mm.443041
- Species
- Mus musculus
- Gene Symbol
- Gm3558
- Entrez Gene ID
- 100041874
- Gene Name
- predicted gene 3558
- Gene Aliases
- ENSMUSG00000079364
- Chromosome Location
- Chr. 14: 7536187 - 7568566 on Build GRCm38
- UniGene ID
- Mm.350858
- Species
- Mus musculus
- Gene Symbol
- Gm3636
- Entrez Gene ID
- 100041151
- Gene Name
- predicted gene 3636
- Gene Aliases
- 100042043
- Chromosome Location
- Chr. 14: 6736742 - 6757604 on Build GRCm38
- UniGene ID
- Mm.396155
- Species
- Mus musculus
- Gene Symbol
- Gm3667
- Entrez Gene ID
- 100042100
- Gene Name
- predicted gene 3667
- Gene Aliases
- Chromosome Location
- Chr. 14: 6869638 - 6892336 on Build GRCm38
- UniGene ID
- Mm.396155
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651813.1 |
- |
1 |
112 |
|
- Gene Symbol
- Gm3696
- Entrez Gene ID
- 100042149
- Gene Name
- predicted gene 3696
- Gene Aliases
- 544988, ENSMUSG00000056940, Gm10014
- Chromosome Location
- Chr. 14: 7056602 - 7105980 on Build GRCm38
- UniGene ID
- Mm.396155
- Species
- Mus musculus
- Gene Symbol
- Gm3715
- Entrez Gene ID
- 100042192
- Gene Name
- predicted gene 3715
- Gene Aliases
- 5033421C21Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.429322
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017182.1 |
- |
|
257 |
|
- Gene Symbol
- Gm3739
- Entrez Gene ID
- 100042235
- Gene Name
- predicted gene 3739
- Gene Aliases
- Chromosome Location
- Chr. 14: 7294750 - 7316522 on Build GRCm38
- UniGene ID
- Mm.483675
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF651817.1 |
- |
1 |
112 |
|
- Gene Symbol
- Gm5458
- Entrez Gene ID
- 432825
- Gene Name
- predicted gene 5458
- Gene Aliases
- EG432825
- Chromosome Location
- Chr. 14: 19594138 - 19602587 on Build GRCm38
- UniGene ID
- Mm.327147
- Species
- Mus musculus
- Gene Symbol
- Gm5795
- Entrez Gene ID
- 544990
- Gene Name
- predicted gene 5795
- Gene Aliases
- Chromosome Location
- Chr. 14: 3188284 - 3196683 on Build GRCm38
- UniGene ID
- Mm.360540
- Species
- Mus musculus
- Gene Symbol
- Gm5796
- Entrez Gene ID
- 545007
- Gene Name
- predicted gene 5796
- Gene Aliases
- Chromosome Location
- Chr. 14: 4018034 - 4076004 on Build GRCm38
- UniGene ID
- Mm.449725
- Species
- Mus musculus
- Gene Symbol
- Gm5797
- Entrez Gene ID
- 545013
- Gene Name
- predicted gene 5797
- Gene Aliases
- EG545013
- Chromosome Location
- Chr. 14: 7323988 - 7332395 on Build GRCm38
- UniGene ID
- Mm.422841
- Species
- Mus musculus
- Gene Symbol
- LOC100036572
- Entrez Gene ID
- 100036572
- Gene Name
- uncharacterized LOC100036572
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.443041
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027416.1 |
- |
|
1276 |
|
- Gene Symbol
- LOC100861615
- Entrez Gene ID
- 100861615
- Gene Name
- alpha takusan-like
- Gene Aliases
- Chromosome Location
- Chr. 14: 6249637 - 6425602 on Build GRCm38
- UniGene ID
- Mm.488431
- Species
- Mus musculus
- Gene Symbol
- LOC545005
- Entrez Gene ID
- 545005
- Gene Name
- uncharacterized LOC545005
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC089308.1 |
- |
1 |
1063 |
|
View Full Product Details