Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243280.1 | NP_001230209.1 | 2 | 804 | |
NM_001243281.1 | NP_001230210.1 | 2 | 804 | ||
NM_001243283.1 | NP_001230212.1 | 2 | 804 | ||
NM_001627.3 | NP_001618.2 | 2 | 804 | ||
GenBank mRNA | AK054632.1 | - | 2 | 757 | |
AK300362.1 | - | 2 | 180 | ||
AK301804.1 | - | 3 | 237 | ||
AK316113.1 | - | 2 | 608 | ||
AL833702.1 | - | 2 | 605 | ||
AY644765.1 | - | 2 | 456 | ||
BC057809.1 | - | 2 | 608 | ||
BC137096.1 | - | 2 | 232 | 9021093 | |
BC137097.1 | - | 2 | 232 | 9021094 | |
DQ486139.1 | - | 2 | 170 | ||
DQ486140.1 | - | 2 | 170 | ||
L38608.1 | - | 2 | 171 | ||
Y10183.1 | - | 2 | 105 |