Gene Transcripts

Gene Symbol
Olr659
Entrez Gene ID
295865
Gene Name
olfactory receptor 659
Gene Aliases
Chromosome Location
Chr. 3: 77414999 - 77415928 on Build Rnor_6.0
UniGene ID
Rn.142842
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000345.1 NP_001000345.1 1 123

View Full Product Details