Gene Transcripts

Gene Symbol
Ddx4
Entrez Gene ID
310090
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases
RVLG
Chromosome Location
Chr. 2: 44447610 - 44504146 on Build Rnor_6.0
UniGene ID
Rn.198577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077647.1 NP_001071115.1 15 1349
GenBank mRNA GQ243745.1 - 14 1129
JQ013737.1 - 14 1137
S75275.1 - 15 1349

View Full Product Details