Gene Transcripts

Gene Symbol
Rnf150
Entrez Gene ID
364983
Gene Name
ring finger protein 150
Gene Aliases
RGD1304572
Chromosome Location
Chr. 19: 24044210 - 24266025 on Build Rnor_6.0
UniGene ID
Rn.205888
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191093.1 NP_001178022.1 6, 7 1201
XM_008772417.1 XP_008770639.1 5, 6 1203

View Full Product Details