Gene Transcripts
- Gene Symbol
- RGD1561827
- Entrez Gene ID
- 502960
- Gene Name
- similar to Interferon alpha-1 precursor
- Gene Aliases
- Chromosome Location
- Chr. 5: 107048349 - 107048927 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_578465.2 |
XP_578465.1
|
|
441 |
|
- Gene Symbol
- LOC100909643
- Entrez Gene ID
- 100909643
- Gene Name
- interferon alpha-1-like
- Gene Aliases
- Chromosome Location
- Chr. 5: 107382726 - 107383824 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100911527
- Entrez Gene ID
- 100911527
- Gene Name
- interferon alpha-12-like
- Gene Aliases
- Chromosome Location
- Chr. 5: 87328395 - 87328946 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1560539
- Entrez Gene ID
- 502966
- Gene Name
- similar to interferon alpha 8/6 precursor; IFNa8/6 (B6)
- Gene Aliases
- Chromosome Location
- Chr. 5: 87353959 - 87354663 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1564637
- Entrez Gene ID
- 502963
- Gene Name
- similar to interferon alpha 8/6 precursor; IFNa8/6 (B6)
- Gene Aliases
- Chromosome Location
- Chr. 5: 107428902 - 107429480 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_578468.3 |
XP_578468.2
|
1 |
441 |
|
View Full Product Details