Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138584.2 | NP_613050.1 | 8 | 1035 | |
GenBank mRNA | AK009332.1 | - | 8 | 1027 | |
AK011985.1 | - | 8 | 1002 | ||
AK090017.1 | - | 8 | 1023 | ||
AK149930.1 | - | 8 | 1009 | ||
AK150215.1 | - | 8 | 1036 | ||
AK150581.1 | - | 8 | 1013 | ||
AK153284.1 | - | 8 | 997 | ||
AK170666.1 | - | 8 | 1028 | ||
AK219439.1 | - | 1 | 58 | ||
BC099434.1 | - | 7 | 800 | 30714184 | |
BC132377.1 | - | 8 | 1012 | 40130654 | |
BC132403.1 | - | 8 | 1012 | 40130680 |