Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159572.1 | NP_001153044.1 | 5, 6 | 877 | |
NM_028732.4 | NP_083008.1 | 6 | 880 | ||
GenBank mRNA | AK014600.1 | - | 6 | 808 | |
AK030479.1 | - | 6 | 820 | ||
AK032383.1 | - | 6 | 825 | ||
AK033393.1 | - | 6 | 826 | ||
AK079734.1 | - | 6 | 828 | ||
AK131118.1 | - | 5, 6 | 793 | ||
AK159086.1 | - | 6 | 825 | ||
AK159864.1 | - | 6 | 825 | ||
AK162635.1 | - | 716 | |||
AK163118.1 | - | 6 | 825 | ||
AK170059.1 | - | 6 | 828 | ||
AK170419.1 | - | 6 | 825 | ||
BC003967.1 | - | 6 | 796 | 3488482 | |
EF426799.1 | - | 6 | 709 | ||
JN602184.1 | - | 6 | 709 |