Gene Transcripts

Gene Symbol
Cyp2b12
Entrez Gene ID
29295
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 12
Gene Aliases
Cyp2b15
Chromosome Location
Chr. 1: 83511167 - 83522965 on Build Rnor_6.0
UniGene ID
Rn.144570
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017156.1 NP_058852.1 787
GenBank mRNA X63545.1 - 787

Gene Symbol
Cyp2b1
Entrez Gene ID
24300
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 1
Gene Aliases
Cyp2b-1, Cypbe
Chromosome Location
Chr. 7: 99142431 - 99183540 on Build Rnor_6.0
UniGene ID
Rn.228579
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134844.1 NP_001128316.1 762
XM_008765491.1 XP_008763713.1 781
XM_008765492.1 XP_008763714.1 781
XM_008765493.1 XP_008763715.1 1143
GenBank mRNA BC166502.1 - 762
J00719.1 - 730
M37134.1 - 730

Gene Symbol
Cyp2b2
Entrez Gene ID
361523
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 2
Gene Aliases
CYPIIB2, Cype
Chromosome Location
Chr. 1: 83103925 - 83119578 on Build Rnor_6.0
UniGene ID
Rn.228579
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198676.1 NP_001185605.1 5 745
GenBank mRNA K01721.1 - 2 244
M34452.1 - 4 434

View Full Product Details