Gene Transcripts

Gene Symbol
LOXHD1
Entrez Gene ID
125336
Gene Name
lipoxygenase homology domains 1
Gene Aliases
DFNB77, LH2D1
Chromosome Location
Chr. 18: 46476961 - 46657115 on Build GRCh38
UniGene ID
Hs.345877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144612.6 NP_653213.6 16 2104
XM_011525804.2 XP_011524106.1 4 338
XM_017025540.1 XP_016881029.1 8 1134
XM_017025541.1 XP_016881030.1 9 1221
XM_017025542.1 XP_016881031.1 9 1221
XM_017025543.1 XP_016881032.1 9 1221
XM_017025544.1 XP_016881033.1 9 1221
XM_017025545.1 XP_016881034.1 9 1221
XM_017025546.1 XP_016881035.1 9 1221
XM_017025547.1 XP_016881036.1 9 1221
XM_017025548.1 XP_016881037.1 16 2186
GenBank mRNA AK127869.1 - 9 1417

View Full Product Details