Gene Transcripts

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
Chromosome Location
Chr. 2: 25232961 - 25342590 on Build GRCh38
UniGene ID
Hs.515840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320892.1 NP_001307821.1 2 165
NM_022552.4 NP_072046.2 2 165
NM_175629.2 NP_783328.1 2 236
NM_175630.1 NP_783329.1 2 236
NR_135490.1 2 236
XM_005264175.4 XP_005264232.1 2 99
XM_011532663.2 XP_011530965.1 2 175
XM_011532664.2 XP_011530966.1 2 176
XM_017003526.1 XP_016859015.1 2 207
GenBank mRNA AB208833.1 - 2 135
AF067972.2 - 2 127
AF331856.1 - 2 115
AL543971.3 - 2 165
BC032392.1 - 2 236
BC043617.1 - 2 135 6150112
BC051864.1 - 2 104

View Full Product Details