Gene Transcripts

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
Chromosome Location
Chr. 3: 184249657 - 184293031 on Build GRCh38
UniGene ID
Hs.146161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032331.3 NP_115707.2 3 598
GenBank mRNA BC005835.2 - 3 594
BC012449.1 - 3 571
BC069005.1 - 3 584

View Full Product Details