Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127219.1 | NP_001120691.1 | 2 | 352 | |
NM_032592.3 | NP_115981.1 | 2 | 648 | ||
XM_005253170.3 | XP_005253227.2 | 2 | 791 | ||
XM_011520406.2 | XP_011518708.1 | 2 | 794 | ||
XM_011520410.2 | XP_011518712.1 | 2 | 792 | ||
XM_017018421.1 | XP_016873910.1 | 2 | 795 | ||
GenBank mRNA | AK057649.1 | - | 2 | 642 | |
AK304074.1 | - | 2 | 585 | ||
AY026508.1 | - | 2 | 603 | ||
BC020197.1 | - | 2 | 622 | 3632065 | |
DA196370.1 | - | 2 | 352 | ||
KU178811.1 | - | 1 | 208 | ||
KU178812.1 | - | 1 | 208 | ||
KU178813.1 | - | 1 | 208 |