Gene Transcripts
- Gene Symbol
- CNTROB
- Entrez Gene ID
- 116840
- Gene Name
- centrobin, centriole duplication and spindle assembly protein
- Gene Aliases
- LIP8, PP1221
- Chromosome Location
- Chr. 17: 7932054 - 7949919 on Build GRCh38
- UniGene ID
- Hs.348012
- Species
- Homo sapiens
View Full Product Details