Gene Transcripts

Gene Symbol
DMPK
Entrez Gene ID
1760
Gene Name
dystrophia myotonica protein kinase
Gene Aliases
DM, DM1, DM1PK, DMK, MDPK, MT-PK
Chromosome Location
Chr. 19: 45769709 - 45782557 on Build GRCh38
UniGene ID
Hs.631596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081560.2 NP_001075029.1 8 1310
NM_001081562.2 NP_001075031.1 8 1310
NM_001081563.2 NP_001075032.1 7 1679
NM_001288764.1 NP_001275693.1 9 1369
NM_001288765.1 NP_001275694.1 6 950
NM_001288766.1 NP_001275695.1 8 1310
NM_004409.4 NP_004400.4 8 1310
GenBank mRNA AB209363.1 - 8 2153
AK297299.1 - 6 901
AK302874.1 - 8 1243
BC026328.1 - 9 1364 4792104
BC062553.1 - 8 1231 6480902
KJ534826.1 - 7 1135
KJ534827.1 - 7 1135
KJ534828.1 - 7 1135
L19268.1 - 8 1232
M87312.1 - 7 1680
M94203.1 - 8 982
S72883.1 - 8 1141

View Full Product Details