Gene Transcripts

Gene Symbol
LOXHD1
Entrez Gene ID
125336
Gene Name
lipoxygenase homology domains 1
Gene Aliases
DFNB77, LH2D1
Chromosome Location
Chr. 18: 46476961 - 46657115 on Build GRCh38
UniGene ID
Hs.345877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144612.6 NP_653213.6 5 590
XM_017025548.1 XP_016881037.1 5 672
GenBank mRNA DR731345.1 - 5 496

View Full Product Details