Gene Transcripts

Gene Symbol
GP5
Entrez Gene ID
2814
Gene Name
glycoprotein V platelet
Gene Aliases
CD42d, GPV
Chromosome Location
Chr. 3: 194394821 - 194399266 on Build GRCh38
UniGene ID
Hs.73734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004488.2 NP_004479.1 2 1656
GenBank mRNA L11238.1 - 1 1697

View Full Product Details