Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009655.2 | NP_033785.1 | 11 | 1786 | |
GenBank mRNA | AB003026.1 | - | 11 | 1448 | |
AK030851.1 | - | 11 | 1787 | ||
AK031391.1 | - | 2862 | |||
BC027280.1 | - | 11 | 1443 | 3501215 | |
L25274.1 | - | 6 | 578 | ||
U95030.1 | - | 11 | 1254 |