Gene Transcripts

Gene Symbol
Dhcr7
Entrez Gene ID
13360
Gene Name
7-dehydrocholesterol reductase
Gene Aliases
AA409147
Chromosome Location
Chr. 7: 143823167 - 143848410 on Build GRCm38
UniGene ID
Mm.249342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007856.2 NP_031882.1 6, 7 1129
GenBank mRNA AF057368.1 - 5, 6 661
AK146811.1 - 5, 6 652
BC006854.1 - 4, 5 643 3157987
CT010277.1 - 4, 5 622

View Full Product Details