Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009088.3 | NP_033114.3 | 20 | 2996 | |
GenBank mRNA | AF000938.1 | - | 20 | 2886 | |
AK031689.1 | - | 21 | 3069 | ||
AK082883.1 | - | 20 | 2979 | ||
AK087773.1 | - | 20 | 3092 | ||
AK147379.1 | - | 20 | 2989 | ||
AK160785.1 | - | 20 | 2989 | ||
AK160970.1 | - | 20 | 2997 | ||
AK178488.1 | - | 2 | 188 | ||
BC010254.1 | - | 20 | 2967 | ||
BC034785.1 | - | 20 | 2981 | ||
BC049190.1 | - | 20 | 2961 | ||
BC053744.1 | - | 20 | 2976 | 6505567 |