Gene Transcripts
- Gene Symbol
- Spg21
- Entrez Gene ID
- 27965
- Gene Name
- spastic paraplegia 21 homolog (human)
- Gene Aliases
- ACP33, BM-019, C78576, D9Wsu18e, GL010, MAST
- Chromosome Location
- Chr. 9: 65460937 - 65488470 on Build GRCm38
- UniGene ID
- Mm.272475
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138584.2 |
NP_613050.1
|
4, 5 |
550 |
|
GenBank mRNA |
AK009332.1 |
- |
4, 5 |
542 |
|
AK011985.1 |
- |
4, 5 |
517 |
|
AK090017.1 |
- |
4, 5 |
538 |
|
AK149930.1 |
- |
4, 5 |
524 |
|
AK150215.1 |
- |
4, 5 |
551 |
|
AK150581.1 |
- |
4, 5 |
528 |
|
AK153284.1 |
- |
4, 5 |
512 |
|
AK170666.1 |
- |
4, 5 |
543 |
|
AK187235.1 |
- |
1, 2 |
95 |
|
AK199934.1 |
- |
1, 2 |
95 |
|
BC099434.1 |
- |
3, 4 |
315 |
30714184 |
BC132377.1 |
- |
4, 5 |
527 |
40130654 |
BC132403.1 |
- |
4, 5 |
527 |
40130680 |
View Full Product Details