Gene Transcripts

Gene Symbol
4933416I08Rik
Entrez Gene ID
71159
Gene Name
RIKEN cDNA 4933416I08 gene
Gene Aliases
Chromosome Location
Chr. X: 53577073 - 53580003 on Build GRCm38
UniGene ID
Mm.396063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027700.1 NP_081976.1 336
GenBank mRNA AK016833.1 - 303
AK163858.1 - 152
CN831548.1 - 2 339

View Full Product Details