Gene Transcripts

Gene Symbol
Kank1
Entrez Gene ID
107351
Gene Name
KN motif and ankyrin repeat domains 1
Gene Aliases
A930031B09Rik, AU015049, AW121052, Ankrd15, D330024H06Rik, mKIAA0172
Chromosome Location
Chr. 19: 25236732 - 25434498 on Build GRCm38
UniGene ID
Mm.260722
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181404.5 NP_852069.4 8 3674
GenBank mRNA AK084638.1 - 1 124
AK122214.1 - 4 2040
AK147945.1 - 5 3261
BC050250.1 - 3 481
BC079563.1 - 4 3001 30535512

View Full Product Details