Gene Transcripts

Gene Symbol
Sprr2j-ps
Entrez Gene ID
20764
Gene Name
small proline-rich protein 2J, pseudogene
Gene Aliases
Sprr2j
Chromosome Location
Chr. 3: 92418087 - 92419396 on Build GRCm38
UniGene ID
Mm.57084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK019074.1 - 2 225
AY158994.1 - 2 228
BC125542.1 - 2 199
BC125544.1 - 2 199

View Full Product Details