Gene Transcripts

Gene Symbol
Med30
Entrez Gene ID
69790
Gene Name
mediator complex subunit 30
Gene Aliases
1810038N03Rik, 2510044J04Rik, TRAP25, Thrap6
Chromosome Location
Chr. 15: 52712445 - 52730431 on Build GRCm38
UniGene ID
Mm.195562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027212.2 NP_081488.1 1, 2 250
GenBank mRNA AK007739.1 - 1, 2 243
AK011104.1 - 1, 2 251
AK146250.1 - 1, 2 255
BC020113.1 - 1, 2 238 4015173

View Full Product Details