Gene Transcripts

Gene Symbol
MINK1
Entrez Gene ID
50488
Gene Name
misshapen like kinase 1
Gene Aliases
B55, MAP4K6, MINK, YSK2, ZC3
Chromosome Location
Chr. 17: 4833340 - 4898061 on Build GRCh38
UniGene ID
Hs.443417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024937.3 NP_001020108.1 4 441
NM_001321236.1 NP_001308165.1 5 557
NM_015716.4 NP_056531.1 4 441
NM_153827.4 NP_722549.2 4 441
NM_170663.4 NP_733763.1 4 441
XM_006721531.2 XP_006721594.1 4 441
XM_006721532.2 XP_006721595.1 4 441
XM_006721536.2 XP_006721599.1 4 441
XM_011523906.1 XP_011522208.1 4 441
XM_017024704.1 XP_016880193.1 4 441
XM_017024705.1 XP_016880194.1 4 441
XM_017024706.1 XP_016880195.1 4 441
XM_017024707.1 XP_016880196.1 4 242
XM_017024708.1 XP_016880197.1 2 92
GenBank mRNA AB035698.1 - 4 197
AB041926.1 - 4 381
AB209450.1 - 4 382
AY775058.1 - 4 197
BC034673.1 - 4 393 4384442
CA392419.1 - 4 414
DA653105.1 - 4 441

View Full Product Details