Gene Transcripts

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
Chromosome Location
Chr. 11: 117205071 - 117232525 on Build GRCh38
UniGene ID
Hs.648612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004716.3 NP_004707.2 17 3361
XM_006718940.3 XP_006719003.1 10 2120
XM_017018546.1 XP_016874035.1 12 1889
GenBank mRNA AK097207.1 - 10 2618
AK127262.1 - 1 4476
BC006357.2 - 16 2733 4125594
BC010696.1 - 17 2931 3868257
U33849.1 - 17 2880
U40623.1 - 15 2751

View Full Product Details