Gene Transcripts

Gene Symbol
BTC
Entrez Gene ID
685
Gene Name
betacellulin
Gene Aliases
Chromosome Location
Chr. 4: 74744759 - 74794718 on Build GRCh38
UniGene ID
Hs.591704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316963.1 NP_001303892.1 5 1031
NM_001729.3 NP_001720.1 6 1178
GenBank mRNA BC011618.2 - 6 969
S55606.1 - 6 1112

View Full Product Details