Gene Transcripts

Gene Symbol
PRSS27
Entrez Gene ID
83886
Gene Name
protease, serine 27
Gene Aliases
CAPH2, MPN
Chromosome Location
Chr. 16: 2712418 - 2720551 on Build GRCh38
UniGene ID
Hs.332878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318395.1 NP_001305324.1 4 836
NM_031948.4 NP_114154.1 5 999
XM_011522689.2 XP_011520991.1 3 728
GenBank mRNA AB056161.1 - 5 658
AJ306593.1 - 5 627
AK055576.1 - 2 1600
AK310214.1 - 4 511
AY030095.1 - 5 608
AY359106.1 - 5 643
BC034294.2 - 5 663 4752547
BC036912.2 - 4 847

View Full Product Details