Gene Transcripts

Gene Symbol
SLC5A5
Entrez Gene ID
6528
Gene Name
solute carrier family 5 member 5
Gene Aliases
NIS, TDH1
Chromosome Location
Chr. 19: 17871394 - 17895175 on Build GRCh38
UniGene ID
Hs.584804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000453.2 NP_000444.1 15 2410
XM_011528192.2 XP_011526494.1 15 3022
XM_011528193.2 XP_011526495.1 16 2525
XM_011528194.2 XP_011526496.1 15 1930
XM_017027158.1 XP_016882647.1 16 2490
GenBank mRNA AF260700.1 - 3 482
BX648217.1 - 12 1624
U66088.1 - 15 2410

View Full Product Details