Gene Transcripts

Gene Symbol
GTF2F1
Entrez Gene ID
2962
Gene Name
general transcription factor IIF subunit 1
Gene Aliases
BTF4, RAP74, TF2F1, TFIIF
Chromosome Location
Chr. 19: 6379569 - 6393280 on Build GRCh38
UniGene ID
Hs.68257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002096.2 NP_002087.2 5 638
GenBank mRNA AK315240.1 - 5 522
BC000120.1 - 5 500 3352627
BC013007.2 - 5 500 3009685
BT007097.1 - 4 353
CR456798.1 - 4 353
X64002.1 - 5 416
X64037.1 - 5 531

View Full Product Details