Gene Transcripts

Gene Symbol
POU4F3
Entrez Gene ID
5459
Gene Name
POU class 4 homeobox 3
Gene Aliases
BRN3C, DFNA15
Chromosome Location
Chr. 5: 146339024 - 146340520 on Build GRCh38
UniGene ID
Hs.553499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002700.2 NP_002691.1 2 967
GenBank mRNA BC104923.1 - 2 967
BC112207.1 - 2 967 8327675

View Full Product Details