Gene Transcripts

Gene Symbol
SLC28A2
Entrez Gene ID
9153
Gene Name
solute carrier family 28 member 2
Gene Aliases
CNT2, HCNT2, HsT17153, SPNT1
Chromosome Location
Chr. 15: 45252230 - 45275934 on Build GRCh38
UniGene ID
Hs.367833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004212.3 NP_004203.2 8, 9 853
XM_011522198.2 XP_011520500.1 8, 9 873
XM_011522199.1 XP_011520501.1 8, 9 824
XM_011522200.2 XP_011520502.1 8, 9 824
XM_011522201.2 XP_011520503.1 8, 9 824
GenBank mRNA AF036109.1 - 7, 8 802
AK222709.1 - 8, 9 853
AK291974.1 - 8, 9 824
BC093737.1 - 7, 8 816 7939582
BC112047.1 - 7, 8 828 8327515
U84392.1 - 8, 9 847

View Full Product Details